siRNA Details
Gene Symbol: Trpv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053686.1 | NP_446138.1 | 2 | 477 | ||
XM_006236375.2 | XP_006236437.1 | 2 | 93 | |||
GenBank mRNA | AF160798.1 | - | 2 | 477 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trpv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053686.1 | NP_446138.1 | 2 | 477 | ||
XM_006236375.2 | XP_006236437.1 | 2 | 93 | |||
GenBank mRNA | AF160798.1 | - | 2 | 477 |