siRNA Details
| Gene Symbol: Stag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053730.1 | NP_446182.1 | 3 | 282 | ||
| XM_006249020.2 | XP_006249082.1 | 3 | 450 | |||
| GenBank mRNA | AY027880.1 | - | 3 | 282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stag3| Gene Symbol: Stag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053730.1 | NP_446182.1 | 3 | 282 | ||
| XM_006249020.2 | XP_006249082.1 | 3 | 450 | |||
| GenBank mRNA | AY027880.1 | - | 3 | 282 | ||