siRNA Details
| Gene Symbol: Cyp27b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053763.1 | NP_446215.1 | 5 | 913 | ||
| XM_008765374.1 | XP_008763596.1 | 6 | 2785 | |||
| GenBank mRNA | AB001992.1 | - | 5 | 913 | ||
| AF000139.1 | - | 5 | 905 | |||
| Gene Symbol: LOC100909462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773774.1 | XP_008771996.1 | 1673 | |||

Cyp27b1
Genomic Map