siRNA Details
| Gene Symbol: Usp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053774.2 | NP_446226.2 | 2 | 273 | ||
| XM_006242861.2 | XP_006242923.1 | 2 | 434 | |||
| GenBank mRNA | AF106659.1 | - | 2 | 273 | ||
| AF202453.1 | - | 1 | 146 | |||
| BC085719.1 | - | 2 | 247 | 7193298 | ||

Usp2
Genomic Map