siRNA Details
Gene Symbol: Ipo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053778.1 | NP_446230.1 | 2 | 822 | ||
GenBank mRNA | AF110195.1 | - | 2 | 822 | ||
BC100658.1 | - | 2 | 830 | 7935813 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ipo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053778.1 | NP_446230.1 | 2 | 822 | ||
GenBank mRNA | AF110195.1 | - | 2 | 822 | ||
BC100658.1 | - | 2 | 830 | 7935813 |