siRNA Details
| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053787.2 | NP_446239.2 | 2 | 317 | ||
| GenBank mRNA | AB032019.1 | - | 2 | 317 | ||
| AF209196.1 | - | 268 | ||||
| AY762624.1 | - | 2 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpv5| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053787.2 | NP_446239.2 | 2 | 317 | ||
| GenBank mRNA | AB032019.1 | - | 2 | 317 | ||
| AF209196.1 | - | 268 | ||||
| AY762624.1 | - | 2 | 152 | |||