siRNA Details
| Gene Symbol: Ift172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053792.1 | NP_446244.1 | 3 | 287 | ||
| GenBank mRNA | AF226993.1 | - | 3 | 287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift172| Gene Symbol: Ift172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053792.1 | NP_446244.1 | 3 | 287 | ||
| GenBank mRNA | AF226993.1 | - | 3 | 287 | ||