siRNA Details
| Gene Symbol: Cited2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053698.2 | NP_446150.1 | 2 | 914 | ||
| GenBank mRNA | AF361476.1 | - | 2 | 919 | ||
| BC087005.1 | - | 2 | 914 | 7191504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cited2| Gene Symbol: Cited2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053698.2 | NP_446150.1 | 2 | 914 | ||
| GenBank mRNA | AF361476.1 | - | 2 | 919 | ||
| BC087005.1 | - | 2 | 914 | 7191504 | ||