siRNA Details
Gene Symbol: Cacna1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053701.1 | NP_446153.1 | 4 | 424 | ||
GenBank mRNA | AF365975.1 | - | 4 | 424 | ||
DQ393415.1 | - | 4 | 428 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacna1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053701.1 | NP_446153.1 | 4 | 424 | ||
GenBank mRNA | AF365975.1 | - | 4 | 424 | ||
DQ393415.1 | - | 4 | 428 |