siRNA Details
| Gene Symbol: Casp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053736.2 | NP_446188.2 | 2 | 298 | ||
| GenBank mRNA | AY029283.1 | - | 2 | 321 | ||
| BC104720.1 | - | 2 | 298 | 7382967 | ||
| FQ227402.1 | - | 2 | 335 | |||
| FQ232279.1 | - | 2 | 335 | |||

Casp4
Genomic Map