siRNA Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053752.2 | NP_446204.2 | 3, 4 | 335 | ||
| GenBank mRNA | BC061537.1 | - | 3, 4 | 339 | 6920009 | |
| FQ212668.1 | - | 3, 4 | 332 | |||
| FQ214532.1 | - | 3, 4 | 294 | |||
| FQ214959.1 | - | 3, 4 | 324 | |||
| FQ215028.1 | - | 3, 4 | 293 | |||
| FQ215469.1 | - | 3, 4 | 332 | |||
| FQ216893.1 | - | 3, 4 | 324 | |||
| FQ217244.1 | - | 3, 4 | 349 | |||

Suclg1
Genomic Map