siRNA Details
| Gene Symbol: Sorbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053770.1 | NP_446222.1 | 8 | 1176 | ||
| GenBank mRNA | AF026505.1 | - | 8 | 1176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sorbs2| Gene Symbol: Sorbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053770.1 | NP_446222.1 | 8 | 1176 | ||
| GenBank mRNA | AF026505.1 | - | 8 | 1176 | ||