siRNA Details
| Gene Symbol: Ipo13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053778.1 | NP_446230.1 | 2 | 1275 | ||
| GenBank mRNA | AF110195.1 | - | 2 | 1275 | ||
| BC100658.1 | - | 2 | 1283 | 7935813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ipo13| Gene Symbol: Ipo13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053778.1 | NP_446230.1 | 2 | 1275 | ||
| GenBank mRNA | AF110195.1 | - | 2 | 1275 | ||
| BC100658.1 | - | 2 | 1283 | 7935813 | ||