siRNA Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053798.2 | NP_446250.1 | 5, 6 | 531 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_030692, AJ297526, BC117753, AK154759, AK161111, and others.

Sacm1l
Genomic Map