siRNA Details
| Gene Symbol: Sardh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053664.2 | NP_446116.1 | 4, 5 | 837 | ||
| XM_006233731.2 | XP_006233793.1 | 5, 6 | 859 | |||
| XM_006233732.2 | XP_006233794.1 | 5, 6 | 1295 | |||
| XM_006233733.2 | XP_006233795.1 | 5, 6 | 861 | |||
| GenBank mRNA | AF067650.1 | - | 4, 5 | 872 | ||
| FQ218777.1 | - | 4, 5 | 1133 | |||
| L79910.1 | - | 3, 4 | 582 | |||
| U62481.1 | - | 3, 4 | 582 | |||

Sardh
Genomic Map