siRNA Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053671.2 | NP_446123.2 | 3, 4 | 1534 | ||
| XM_008763143.1 | XP_008761365.1 | 3, 4 | 3056 | |||
| GenBank mRNA | AF107843.1 | - | 3, 4 | 1457 | ||
| CK473511.1 | - | 2, 3 | 386 | |||
| Gene Symbol: LOC102548101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762223.1 | XP_008760445.1 | 1 | 317 | ||

Tmf1
Genomic Map