siRNA Details
| Gene Symbol: Dnajc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053690.2 | NP_446142.2 | 3, 4 | 1620 | ||
| XM_006240726.1 | XP_006240788.1 | 3, 4 | 1693 | |||
| XM_006240727.1 | XP_006240789.1 | 2, 3 | 1559 | |||
| XM_008765011.1 | XP_008763233.1 | 3, 4 | 1682 | |||
| GenBank mRNA | AF351783.1 | - | 3, 4 | 1668 | ||
| BC083549.1 | - | 3, 4 | 1620 | 7127738 | ||
| Gene Symbol: LOC498205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769500.1 | XP_008767722.1 | 1 | 1576 | ||

Dnajc14
Genomic Map