siRNA Details
| Gene Symbol: Rasgrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053721.1 | NP_446173.1 | 22 | 2983 | ||
| GenBank mRNA | AJ276774.1 | - | 22 | 2983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasgrf2| Gene Symbol: Rasgrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053721.1 | NP_446173.1 | 22 | 2983 | ||
| GenBank mRNA | AJ276774.1 | - | 22 | 2983 | ||