siRNA Details
| Gene Symbol: Cntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053746.1 | NP_446198.1 | 13 | 1860 | ||
| XM_008765902.1 | XP_008764124.1 | 13 | 1925 | |||
| GenBank mRNA | D87212.1 | - | 13 | 1860 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn5| Gene Symbol: Cntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053746.1 | NP_446198.1 | 13 | 1860 | ||
| XM_008765902.1 | XP_008764124.1 | 13 | 1925 | |||
| GenBank mRNA | D87212.1 | - | 13 | 1860 | ||