siRNA Details
| Gene Symbol: Dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053748.1 | NP_446200.1 | 7, 8 | 935 | ||
| XM_006230633.2 | XP_006230695.1 | 8, 9 | 1122 | |||
| GenBank mRNA | BC107673.1 | - | 8, 9 | 946 | 7938129 | |
| D89340.2 | - | 7, 8 | 935 | |||

Dpp3
Genomic Map