siRNA Details
| Gene Symbol: Aurkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053749.1 | NP_446201.1 | 7 | 786 | ||
| XM_006246569.2 | XP_006246631.1 | 7 | 804 | |||
| XM_006246570.2 | XP_006246632.1 | 6 | 691 | |||
| XM_008767757.1 | XP_008765979.1 | 7 | 701 | |||
| XM_008767758.1 | XP_008765980.1 | 7 | 647 | |||
| GenBank mRNA | BC097297.1 | - | 7 | 704 | 7454285 | |
| D89731.1 | - | 7 | 786 | |||

Aurkb
Genomic Map