siRNA Details
| Gene Symbol: Opcml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053848.1 | NP_446300.1 | 1, 2 | 641 | ||
| GenBank mRNA | M88709.1 | - | 791 | |||
| M88710.1 | - | 1, 2 | 472 | |||
| M88711.1 | - | 1, 2 | 641 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Opcml| Gene Symbol: Opcml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053848.1 | NP_446300.1 | 1, 2 | 641 | ||
| GenBank mRNA | M88709.1 | - | 791 | |||
| M88710.1 | - | 1, 2 | 472 | |||
| M88711.1 | - | 1, 2 | 641 | |||