siRNA Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053865.1 | NP_446317.1 | 5 | 2426 | ||
| XM_008764690.1 | XP_008762912.1 | 3 | 456 | |||
| GenBank mRNA | FQ212949.1 | - | 3 | 367 | ||
| FQ214343.1 | - | 3 | 292 | |||
| U17603.1 | - | 3 | 426 | |||
| U17604.1 | - | 5 | 2426 | |||

Rtn1
Genomic Map