siRNA Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053866.3 | NP_446318.3 | 2 | 386 | ||
| GenBank mRNA | CK366206.1 | - | 2 | 386 | ||
| FQ211509.1 | - | 447 | ||||
| FQ223782.1 | - | 2 | 461 | |||
| U17901.1 | - | 2 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plaa| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053866.3 | NP_446318.3 | 2 | 386 | ||
| GenBank mRNA | CK366206.1 | - | 2 | 386 | ||
| FQ211509.1 | - | 447 | ||||
| FQ223782.1 | - | 2 | 461 | |||
| U17901.1 | - | 2 | 245 | |||