siRNA Details
| Gene Symbol: Rere | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053885.2 | NP_446337.2 | 18 | 3846 | ||
| XM_006239438.2 | XP_006239500.1 | 18 | 3919 | |||
| XM_006239441.2 | XP_006239503.1 | 12 | 5826 | |||
| XM_008764198.1 | XP_008762420.1 | 18 | 3919 | |||
| XM_008764199.1 | XP_008762421.1 | 18 | 4336 | |||
| XM_008764200.1 | XP_008762422.1 | 18 | 3919 | |||
| XM_008764201.1 | XP_008762423.1 | 17 | 3794 | |||
| XM_008764202.1 | XP_008762424.1 | 17 | 3360 | |||
| XM_008764203.1 | XP_008762425.1 | 16 | 5487 | |||
| GenBank mRNA | U44091.1 | - | 17 | 4655 | ||

Rere
Genomic Map