siRNA Details
| Gene Symbol: Myt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053888.1 | NP_446340.1 | 9 | 1097 | ||
| GenBank mRNA | U48809.1 | - | 9 | 1117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myt1l| Gene Symbol: Myt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053888.1 | NP_446340.1 | 9 | 1097 | ||
| GenBank mRNA | U48809.1 | - | 9 | 1117 | ||