siRNA Details
| Gene Symbol: Dnase1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053907.1 | NP_446359.1 | 1 | 290 | ||
| GenBank mRNA | AF039852.1 | - | 1 | 240 | ||
| BC088122.1 | - | 1 | 288 | 7325326 | ||
| FQ226037.1 | - | 1 | 117 | |||
| FQ227612.1 | - | 1 | 117 | |||
| FQ230283.1 | - | 1 | 117 | |||
| FQ230990.1 | - | 1 | 117 | |||
| FQ231646.1 | - | 1 | 116 | |||
| FQ231744.1 | - | 1 | 116 | |||
| FQ231870.1 | - | 1 | 120 | |||
| FQ231939.1 | - | 1 | 117 | |||
| FQ231976.1 | - | 1 | 173 | |||
| FQ233228.1 | - | 1 | 173 | |||
| FQ234709.1 | - | 1 | 173 | |||
| U75689.1 | - | 1 | 290 | |||

Dnase1l3
Genomic Map