siRNA Details
| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053908.1 | NP_446360.1 | 3 | 337 | ||
| GenBank mRNA | BC099824.1 | - | 3 | 194 | 7932620 | |
| U77038.1 | - | 3 | 337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn6| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053908.1 | NP_446360.1 | 3 | 337 | ||
| GenBank mRNA | BC099824.1 | - | 3 | 194 | 7932620 | |
| U77038.1 | - | 3 | 337 | |||