siRNA Details
| Gene Symbol: CIDEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318807.1 | NP_001305736.1 | 6, 7 | 1628 | ||
| NM_014430.2 | NP_055245.2 | 5, 6 | 1479 | |||
| XM_011536659.2 | XP_011534961.1 | 5, 6 | 1135 | |||
| XM_017021221.1 | XP_016876710.1 | 5, 6 | 3183 | |||
| GenBank mRNA | AF190901.1 | - | 5, 6 | 1523 | ||
| AF218586.1 | - | 3, 4 | 657 | |||
| AF544398.1 | - | 3, 4 | 440 | |||
| AK000387.1 | - | 3, 4 | 434 | |||
| AK122828.1 | - | 5, 6 | 3071 | |||
| BC035970.1 | - | 3, 4 | 467 | 4716269 | ||
| CB162472.1 | - | 3, 4 | 415 | |||

CIDEB
Genomic Map