siRNA Details
| Gene Symbol: Cyth2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053911.2 | NP_446363.1 | 4, 5 | 544 | ||
| XM_006228964.2 | XP_006229026.1 | 4, 5 | 562 | |||
| XM_008759304.1 | XP_008757526.1 | 3, 4 | 1035 | |||
| GenBank mRNA | FQ229744.1 | - | 3, 4 | 265 | ||
| U83896.1 | - | 4, 5 | 548 | |||

Cyth2
Genomic Map