siRNA Details
| Gene Symbol: Psmd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053925.1 | NP_446377.1 | 77 | |||
| GenBank mRNA | AB022014.1 | - | 77 | |||
| BC158584.1 | - | 87 | 8369120 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd10| Gene Symbol: Psmd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053925.1 | NP_446377.1 | 77 | |||
| GenBank mRNA | AB022014.1 | - | 77 | |||
| BC158584.1 | - | 87 | 8369120 | |||