siRNA Details
| Gene Symbol: Cltb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053835.1 | NP_446287.1 | 4 | 474 | ||
| XM_006253595.2 | XP_006253657.1 | 4 | 562 | |||
| GenBank mRNA | FQ213983.1 | - | 4 | 564 | ||
| FQ229322.1 | - | 4 | 492 | |||
| M15883.1 | - | 4 | 474 | |||
| M19262.1 | - | 4 | 474 | |||

Cltb
Genomic Map