siRNA Details
| Gene Symbol: Nrxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053846.1 | NP_446298.1 | 5 | 1079 | ||
| XM_008760057.1 | XP_008758279.1 | 6 | 1333 | |||
| XM_008760058.1 | XP_008758280.1 | 6 | 1333 | |||
| XM_008760059.1 | XP_008758281.1 | 6 | 1333 | |||
| XM_008760060.1 | XP_008758282.1 | 6 | 1333 | |||
| XM_008760061.1 | XP_008758283.1 | 6 | 1333 | |||
| XM_008760062.1 | XP_008758284.1 | 6 | 1333 | |||
| XM_008760063.1 | XP_008758285.1 | 6 | 1333 | |||
| GenBank mRNA | M96376.1 | - | 5 | 1079 | ||

Nrxn2
Genomic Map