siRNA Details
| Gene Symbol: Pdia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053849.1 | NP_446301.1 | 3 | 617 | ||
| GenBank mRNA | BC061535.1 | - | 3 | 431 | 6922170 | |
| M86870.1 | - | 3 | 617 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdia4| Gene Symbol: Pdia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053849.1 | NP_446301.1 | 3 | 617 | ||
| GenBank mRNA | BC061535.1 | - | 3 | 431 | 6922170 | |
| M86870.1 | - | 3 | 617 | |||