siRNA Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053866.3 | NP_446318.3 | 2, 3 | 448 | ||
| GenBank mRNA | CK366206.1 | - | 2, 3 | 448 | ||
| CV109083.1 | - | 1, 2 | 21 | |||
| FQ211509.1 | - | 509 | ||||
| FQ223782.1 | - | 2, 3 | 523 | |||
| U17901.1 | - | 2, 3 | 307 | |||

Plaa
Genomic Map