siRNA Details
| Gene Symbol: Cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053879.1 | NP_446331.1 | 5 | 586 | ||
| XM_008763166.1 | XP_008761388.1 | 1 | 317 | |||
| GenBank mRNA | U35371.1 | - | 5 | 586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn4| Gene Symbol: Cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053879.1 | NP_446331.1 | 5 | 586 | ||
| XM_008763166.1 | XP_008761388.1 | 1 | 317 | |||
| GenBank mRNA | U35371.1 | - | 5 | 586 | ||