siRNA Details
| Gene Symbol: UBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128160.1 | NP_001121632.1 | 2 | 682 | ||
| NM_001128161.1 | NP_001121633.1 | 3 | 301 | |||
| NM_014517.4 | NP_055332.3 | 2 | 682 | |||
| GenBank mRNA | AF198487.1 | - | 2 | 326 | ||
| AK023274.1 | - | 2 | 314 | |||
| AK293100.1 | - | 3 | 301 | |||
| BC047235.1 | - | 2 | 681 | 5262788 | ||
| DB131031.1 | - | 3 | 274 | |||

UBP1
Genomic Map