siRNA Details
Gene Symbol: Trip10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053920.1 | NP_446372.1 | 4 | 290 | ||
GenBank mRNA | AB006914.1 | - | 4 | 290 | ||
BC061840.1 | - | 4 | 292 | 5600051 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trip10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053920.1 | NP_446372.1 | 4 | 290 | ||
GenBank mRNA | AB006914.1 | - | 4 | 290 | ||
BC061840.1 | - | 4 | 292 | 5600051 |