siRNA Details
| Gene Symbol: Pex12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053921.1 | NP_446373.1 | 3 | 499 | ||
| XM_006246967.2 | XP_006247029.1 | 2 | 285 | |||
| XM_008767934.1 | XP_008766156.1 | 2 | 290 | |||
| GenBank mRNA | AB002111.1 | - | 3 | 499 | ||
| BC072481.1 | - | 3 | 455 | 7104469 | ||
| Gene Symbol: LOC100909787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750890.3 | XP_003750938.1 | 3 | 499 | ||
| XM_006247026.2 | XP_006247088.1 | 2 | 285 | |||
| XM_008773868.1 | XP_008772090.1 | 2 | 290 | |||

Pex12
Genomic Map