siRNA Details
| Gene Symbol: Acacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053922.1 | NP_446374.1 | 1 | 262 | ||
| GenBank mRNA | AB004329.1 | - | 1 | 262 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acacb| Gene Symbol: Acacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053922.1 | NP_446374.1 | 1 | 262 | ||
| GenBank mRNA | AB004329.1 | - | 1 | 262 | ||