siRNA Details
| Gene Symbol: Pik3c2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053923.1 | NP_446375.1 | 2 | 228 | ||
| XM_008763348.1 | XP_008761570.1 | 2 | 280 | |||
| GenBank mRNA | AB009636.1 | - | 2 | 228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3c2g| Gene Symbol: Pik3c2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053923.1 | NP_446375.1 | 2 | 228 | ||
| XM_008763348.1 | XP_008761570.1 | 2 | 280 | |||
| GenBank mRNA | AB009636.1 | - | 2 | 228 | ||