siRNA Details
| Gene Symbol: Bak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053812.1 | NP_446264.1 | 3 | 346 | ||
| XM_006256101.2 | XP_006256163.1 | 3 | 365 | |||
| GenBank mRNA | AF259504.1 | - | 3 | 346 | ||
| BC089784.1 | - | 3 | 307 | 7366192 | ||

Bak1
Genomic Map