siRNA Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034022.1 | NP_001029194.1 | 8 | 1002 | ||
| NM_053814.2 | NP_446266.2 | 8 | 1002 | |||
| XM_006246422.2 | XP_006246484.1 | 8 | 1624 | |||
| XM_006246423.2 | XP_006246485.1 | 8 | 1624 | |||
| XM_006246424.2 | XP_006246486.1 | 8 | 1624 | |||
| XM_006246425.2 | XP_006246487.1 | 8 | 1269 | |||
| XM_006246427.2 | XP_006246489.1 | 8 | 1270 | |||
| XM_006246430.2 | XP_006246492.1 | 2 | 3399 | |||
| XM_006246431.2 | XP_006246493.1 | 8 | 1611 | |||
| XM_006246432.2 | XP_006246494.1 | 8 | 1255 | |||
| XM_008767759.1 | XP_008765981.1 | 8 | 1624 | |||
| XM_008767760.1 | XP_008765982.1 | 8 | 1624 | |||
| XM_008767761.1 | XP_008765983.1 | 8 | 1625 | |||
| XM_008767762.1 | XP_008765984.1 | 8 | 1611 | |||
| GenBank mRNA | AF311311.1 | - | 8 | 932 | ||
| BC098911.1 | - | 8 | 988 | 7446349 | ||
| FQ222001.1 | - | 8 | 1394 | |||

Mprip
Genomic Map