siRNA Details
| Gene Symbol: Timp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053819.1 | NP_446271.1 | 5 | 454 | ||
| XM_006256608.2 | XP_006256670.1 | 5 | 518 | |||
| XM_008773034.1 | XP_008771256.1 | 4 | 428 | |||
| GenBank mRNA | AF411319.1 | - | 2 | 204 | ||
| AY550026.1 | - | 4 | 443 | |||
| BC099821.1 | - | 5 | 452 | 7930800 | ||
| FQ219856.1 | - | 5 | 464 | |||
| FQ221763.1 | - | 5 | 464 | |||
| FQ222271.1 | - | 5 | 464 | |||
| FQ222437.1 | - | 5 | 467 | |||
| FQ222915.1 | - | 5 | 464 | |||
| FQ228400.1 | - | 5 | 463 | |||
| FQ230039.1 | - | 5 | 461 | |||
| L31883.1 | - | 5 | 454 | |||
| U06179.1 | - | 4 | 437 | |||

Timp1
Genomic Map