siRNA Details
| Gene Symbol: Ralb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053821.2 | NP_446273.1 | 4, 5 | 725 | ||
| GenBank mRNA | BC072505.1 | - | 4, 5 | 725 | 7099019 | |
| L19699.1 | - | 3, 4 | 573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ralb| Gene Symbol: Ralb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053821.2 | NP_446273.1 | 4, 5 | 725 | ||
| GenBank mRNA | BC072505.1 | - | 4, 5 | 725 | 7099019 | |
| L19699.1 | - | 3, 4 | 573 | |||