siRNA Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053851.1 | NP_446303.1 | 11 | 1435 | ||
| XM_006254298.2 | XP_006254360.1 | 12 | 1403 | |||
| XM_006254300.2 | XP_006254362.1 | 11 | 1344 | |||
| XM_006254301.2 | XP_006254363.1 | 11 | 2301 | |||
| XM_006254303.2 | XP_006254365.1 | 11 | 1600 | |||
| XM_006254304.2 | XP_006254366.1 | 11 | 2252 | |||
| GenBank mRNA | AF394941.1 | - | 11 | 1326 | ||
| AF423193.1 | - | 11 | 1258 | |||
| AY190119.1 | - | 12 | 1141 | |||
| AY190120.1 | - | 11 | 1081 | |||
| M80545.1 | - | 11 | 1435 | |||

Cacnb2
Genomic Map