siRNA Details
| Gene Symbol: OR1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014565.2 | NP_055380.2 | 1 | 77 | ||
| GenBank mRNA | BC069169.1 | - | 1 | 77 | 7216916 | |
| BC128248.1 | - | 1 | 77 | |||
| BC141934.1 | - | 1 | 77 | 40114773 | ||
| BC141935.1 | - | 1 | 77 | 40114774 | ||

OR1A1
Genomic Map