siRNA Details
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053888.1 | NP_446340.1 | 10 | 1352 | ||
GenBank mRNA | U48809.1 | - | 10 | 1372 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053888.1 | NP_446340.1 | 10 | 1352 | ||
GenBank mRNA | U48809.1 | - | 10 | 1372 |