siRNA Details
| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053908.1 | NP_446360.1 | 15 | 1912 | ||
| GenBank mRNA | AF468653.1 | - | 8 | 881 | ||
| BC099824.1 | - | 15 | 1769 | 7932620 | ||
| FQ232747.1 | - | 9 | 962 | |||
| U77038.1 | - | 15 | 1912 | |||

Ptpn6
Genomic Map