siRNA Details
| Gene Symbol: Cyth3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053912.2 | NP_446364.2 | 7, 8 | 839 | ||
| GenBank mRNA | BC101884.1 | - | 7, 8 | 839 | 7929513 | |
| U83897.1 | - | 7, 8 | 832 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyth3| Gene Symbol: Cyth3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053912.2 | NP_446364.2 | 7, 8 | 839 | ||
| GenBank mRNA | BC101884.1 | - | 7, 8 | 839 | 7929513 | |
| U83897.1 | - | 7, 8 | 832 | |||